missing translation for 'onlineSavingsMsg'
Learn More

1-Tridecene 97.0+%, TCI America™

Catalog No. T063725ML
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T063725ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T063725ML Supplier TCI America Supplier No. T063725ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2437-56-1
Molecular Formula C13H26
Molecular Weight (g/mol) 182.351
MDL Number MFCD00008976
InChI Key VQOXUMQBYILCKR-UHFFFAOYSA-N
Synonym 1-tridecene, n-tridec-1-ene, tridecylene, alpha-tridecene, undecylethylene, tridecene-1, unii-5b0u2s314c, ccris 5719, 1-tridecylene, .alpha.-tridecene
PubChem CID 17095
IUPAC Name tridec-1-ene
SMILES CCCCCCCCCCCC=C

Specifications

Color Colorless
Boiling Point 233°C
Quantity 25 mL
Formula Weight 182.35
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Tridecene
EINECSNumber (2)-0027
RTECSNumber YD4595000
TSCA Yes