accessibility menu, dialog, popup

UserName

CAS RN 2432-51-1

IUPAC Name: 1-(methylsulfanyl)butan-1-one
Synonyms: 1-(methylsulfanyl)butan-1-one
Molecular Weight (g/mol): 118.19
Molecular Formula: C5H10OS
InChi Key: GRLJIIJNZJVMGP-UHFFFAOYSA-N
SMILES: CCCC(=O)SC
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)