accessibility menu, dialog, popup

UserName

CAS RN 24277-39-2

H3CH3CCH3OONHOHOOOCH3H3CCH3

CAS RN 24277-39-2

IUPAC Name: 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
Synonyms: 5-(tert-butoxy)-4-[(tert-butoxycarbonyl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol): 303.36
Molecular Formula: C14H25NO6
InChi Key: YMOYURYWGUWMFM-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)