missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-glutamic acid 1-tert-butyl ester, 99%
CAS: 24277-39-2 | C14H25NO6 | 303.36 g/mol
$130.27 - $370.71
Chemical Identifiers
| CAS | 24277-39-2 |
|---|---|
| Molecular Formula | C14H25NO6 |
| Molecular Weight (g/mol) | 303.36 |
| MDL Number | MFCD00038273 |
| InChI Key | YMOYURYWGUWMFM-UHFFFAOYNA-N |
| Synonym | boc-glu-otbu, boc-l-glutamic acid 1-tert-butyl ester, s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, n-t-boc-l-glutamic acid a-t-butyl-ester, s-5-tert-butoxy-4-tert-butoxycarbonylamino-5-oxopentanoic acid, 4s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, t-boc-glu-otbu, n-boc-l-glutamic acid 1-tert-butyl ester, pubchem19036, ksc495g0n |
| PubChem CID | 6993444 |
| SMILES | CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH5984203
|
Thermo Scientific Chemicals
H5984203 |
1 g |
Each for $130.27
|
|
|||||
|
AAH5984206
|
Thermo Scientific Chemicals
H5984206 |
5 g |
Each for $370.71
|
|
|||||
Description
Boc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBoc-Glu-OtBu is an N-terminal protected amino acid used in solid-phase peptide synthesis (SPPS) to make unique peptides containing glutamate tert-butyl ester residues.
Solubility
Soluble in dimethyl formamide.
Notes
Recommended storage temperature: -20°C. Store away from oxidizing agents.
Chemical Identifiers
| 24277-39-2 | |
| 303.36 | |
| YMOYURYWGUWMFM-UHFFFAOYNA-N | |
| 6993444 |
| C14H25NO6 | |
| MFCD00038273 | |
| boc-glu-otbu, boc-l-glutamic acid 1-tert-butyl ester, s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, n-t-boc-l-glutamic acid a-t-butyl-ester, s-5-tert-butoxy-4-tert-butoxycarbonylamino-5-oxopentanoic acid, 4s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, t-boc-glu-otbu, n-boc-l-glutamic acid 1-tert-butyl ester, pubchem19036, ksc495g0n | |
| CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C |
Specifications
| 24277-39-2 | |
| MFCD00038273 | |
| 3653769 | |
| Soluble in dimethyl formamide. | |
| CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C | |
| 303.36 | |
| 303.36 | |
| N-Boc-L-glutamic acid 1-tert-butyl ester |
| C14H25NO6 | |
| 1 g | |
| boc-glu-otbu, boc-l-glutamic acid 1-tert-butyl ester, s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, n-t-boc-l-glutamic acid a-t-butyl-ester, s-5-tert-butoxy-4-tert-butoxycarbonylamino-5-oxopentanoic acid, 4s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, t-boc-glu-otbu, n-boc-l-glutamic acid 1-tert-butyl ester, pubchem19036, ksc495g0n | |
| YMOYURYWGUWMFM-UHFFFAOYNA-N | |
| 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid | |
| 6993444 | |
| 99% |
Safety and Handling
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only