missing translation for 'onlineSavingsMsg'
Learn More

1-tert-Butyl N-(tert-Butoxycarbonyl)-L-glutamate 97.0+%, TCI America™

Catalog No. p-7123600
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
B45345G 5 g
B453425G 25 g
2 options

Catalog No. B45345G

Supplier: TCI America B45345G

Only null left

Chemical Identifiers

CAS 24277-39-2
Molecular Formula C14H25NO6
Molecular Weight (g/mol) 303.36
MDL Number MFCD00038273
InChI Key YMOYURYWGUWMFM-UHFFFAOYNA-N
Synonym boc-glu-otbu, boc-l-glutamic acid 1-tert-butyl ester, s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, n-t-boc-l-glutamic acid a-t-butyl-ester, s-5-tert-butoxy-4-tert-butoxycarbonylamino-5-oxopentanoic acid, 4s-5-tert-butoxy-4-tert-butoxycarbonyl amino-5-oxopentanoic acid, t-boc-glu-otbu, n-boc-l-glutamic acid 1-tert-butyl ester, pubchem19036, ksc495g0n
PubChem CID 6993444
IUPAC Name 5-(tert-butoxy)-4-{[(tert-butoxy)carbonyl]amino}-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)NC(CCC(O)=O)C(=O)OC(C)(C)C

Specifications

Melting Point 113°C
Color White
Quantity 5 g
Formula Weight 303.36
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butyl N-(tert-Butoxycarbonyl)-L-glutamate
TSCA No
Recommended Storage Freezer