accessibility menu, dialog, popup

UserName

CAS RN 2424-90-0

HOOOHO

CAS RN 2424-90-0

IUPAC Name: heptadecanedioic acid
Synonyms: heptadecanedioic acid
Molecular Weight (g/mol): 300.44
Molecular Formula: C17H32O4
InChi Key: QCNWZROVPSVEJA-UHFFFAOYSA-N
SMILES: OC(=O)CCCCCCCCCCCCCCCC(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results