missing translation for 'onlineSavingsMsg'
Learn More

Heptadecanedioic Acid 95.0+%, TCI America™

Catalog No. H09721G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09721G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H09721G Supplier TCI America Supplier No. H09721G
Only null left

Chemical Identifiers

CAS 2424-90-0
Molecular Formula C17H32O4
Molecular Weight (g/mol) 300.439
MDL Number MFCD00673439
InChI Key QCNWZROVPSVEJA-UHFFFAOYSA-N
Synonym 1,15-Pentadecanedicarboxylic Acid
PubChem CID 3083765
IUPAC Name heptadecanedioic acid
SMILES C(CCCCCCCC(=O)O)CCCCCCCC(=O)O

Specifications

Melting Point 121°C
Color White
Quantity 1 g
Formula Weight 300.44
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Heptadecanedioic Acid