accessibility menu, dialog, popup

UserName

FFFN

CAS RN 241154-09-6

IUPAC Name: 2,3,5-trifluorobenzonitrile
Synonyms: 2,3,5-trifluorobenzonitrile
Molecular Weight (g/mol): 157.1
Molecular Formula: C7H2F3N
InChi Key: MMPKVFQMDMBXSG-UHFFFAOYSA-N
SMILES: FC1=CC(F)=C(F)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results