accessibility menu, dialog, popup

UserName

CAS RN 24036-52-0

IUPAC Name: 6-bromo-3,4-dihydro-2H-1,4-benzoxazin-3-one
Synonyms: 6-bromo-2,4-dihydro-1,4-benzoxazin-3-one
Molecular Weight (g/mol): 228.05
Molecular Formula: C8H6BrNO2
InChi Key: UQCFMEFQBSYDHY-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(OCC(=O)N2)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)