missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2H-1,4-benzoxazin-3(4H)-one 94.0+%, TCI America™

Catalog No. B44025G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B44025G 5 g
B44021G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B44025G Supplier TCI America Supplier No. B44025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24036-52-0
Molecular Formula C8H6BrNO2
Molecular Weight (g/mol) 228.045
MDL Number MFCD00461173
InChI Key UQCFMEFQBSYDHY-UHFFFAOYSA-N
Synonym 6-bromo-2h-1,4-benzoxazin-3 4h-one, 6-bromo-2h-benzo b 1,4 oxazin-3 4h-one, 6-bromo-3-oxo-4h-benzo 1,4 oxazine, 6-bromo-1,4-benzoxazin-3-one, 6-bromo-3,4-dihydro-2h-1,4-benzoxazin-3-one, 2h-1,4-benzoxazin-3 4h-one, 6-bromo, 6-bromo-2,4-dihydro-1,4-benzoxazin-3-one, 6-bromo-4h-benzo 1,4 oxazin-3-one, pubchem16746
PubChem CID 16218142
IUPAC Name 6-bromo-4H-1,4-benzoxazin-3-one
SMILES C1C(=O)NC2=C(O1)C=CC(=C2)Br

Specifications

Melting Point 221°C
Color Yellow
Quantity 5 g
Formula Weight 228.05
Percent Purity ≥94.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 6-Bromo-2H-1,4-benzoxazin-3(4H)-one