accessibility menu, dialog, popup

UserName

ClClClNCl

CAS RN 2402-79-1

IUPAC Name: 2,3,5,6-tetrachloropyridine
Synonyms: 2,3,5,6-tetrachlorpyridine
Molecular Weight (g/mol): 216.87
Molecular Formula: C5HCl4N
InChi Key: FATBKZJZAHWCSL-UHFFFAOYSA-N
SMILES: ClC1=CC(Cl)=C(Cl)N=C1Cl
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)