missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrachloropyridine 98.0+%, TCI America™

Catalog No. T23315G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T23315G 5 g
T233125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T23315G Supplier TCI America Supplier No. T23315G

Chemical Identifiers

CAS 2402-79-1
Molecular Formula C5HCl4N
Molecular Weight (g/mol) 216.87
MDL Number MFCD00092428
InChI Key FATBKZJZAHWCSL-UHFFFAOYSA-N
Synonym pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine
PubChem CID 16990
IUPAC Name 2,3,5,6-tetrachloropyridine
SMILES ClC1=CC(Cl)=C(Cl)N=C1Cl

Specifications

Melting Point 93°C
Color White
Boiling Point 251°C
Quantity 5 g
Formula Weight 216.87
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,5,6-Tetrachloropyridine
RTECSNumber UT8225000
TSCA Yes