missing translation for 'onlineSavingsMsg'
Learn More

2,3,5,6-Tetrachloropyridine, 98%

Catalog No. AAL1945803
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1945803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL1945803 Supplier Thermo Scientific Chemicals Supplier No. L1945803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2402-79-1 | C5HCl4N | 216.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2402-79-1
Molecular Formula C5HCl4N
Molecular Weight (g/mol) 216.87
MDL Number MFCD00092428
InChI Key FATBKZJZAHWCSL-UHFFFAOYSA-N
Synonym pyridine, 2,3,5,6-tetrachloro, unii-wjr8hay65e, wjr8hay65e, 2,3,5,6-tetrachlorpyridine, tetrachloropyridine, 2,3,5,6, dsstox_cid_7467, acmc-209g7z, 2,5,6-tetrachloropyridine, dsstox_rid_78464, 2,3,5,6tetrachloropyridine
PubChem CID 16990
IUPAC Name 2,3,5,6-tetrachloropyridine
SMILES ClC1=CC(Cl)=C(Cl)N=C1Cl

Specifications

Melting Point 89°C to 93°C
Boiling Point 250°C to 252°C
Flash Point 188°C (370°F)
Quantity 1 g
UN Number UN3077
Beilstein 129639
Formula Weight 216.88
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrachloropyridine
Hazard Category H302-H317
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 219-283-9
RTECSNumber UT8225000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only