accessibility menu, dialog, popup

UserName

OONH2OOH2N

CAS RN 23978-55-4

IUPAC Name: 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium
Synonyms: 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium
Molecular Weight (g/mol): 264.37
Molecular Formula: C12H28N2O4
InChi Key: NLMDJJTUQPXZFG-UHFFFAOYSA-P
SMILES: C1COCCOCC[NH2+]CCOCCOCC[NH2+]1
Percent Purity 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)