missing translation for 'onlineSavingsMsg'
Learn More

1,10-Diaza-18-crown-6, 96%

Catalog No. AAH51063MD
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH51063MD 250 mg
AAH5106303 1 g
AAH5106306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH51063MD Supplier Thermo Scientific Chemicals Supplier No. H51063MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 23978-55-4 | C12H28N2O4 | 264.37 g/mol

1,10-Diaza-18-crown-6 are starting materials to produce macrocyclic polyamine metal-complexing agents. It is used as a ligand in chemistry for the applications of diagnostic imaging technique to identify medical condition or disease and for the development of medication as a therapeutic agent and pharmaceutic aid. Also used as an ether.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 23978-55-4
Molecular Formula C12H28N2O4
Molecular Weight (g/mol) 264.37
MDL Number MFCD00005112
InChI Key NLMDJJTUQPXZFG-UHFFFAOYSA-P
Synonym kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
PubChem CID 72805
IUPAC Name 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane
SMILES C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

Specifications

Quantity 250 mg
Beilstein 609764
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 262.35
Percent Purity 96%
Chemical Name or Material 1,10-Diaza-18-crown-6
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 245-965-0
RTECSNumber HM1650000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only