missing translation for 'onlineSavingsMsg'
Learn More

4,13-Diaza-18-crown 6-Ether 98.0+%, TCI America™

Catalog No. D23235G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23235G 5 g
D23231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23235G Supplier TCI America Supplier No. D23235G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23978-55-4
Molecular Formula C12H28N2O4
Molecular Weight (g/mol) 264.37
MDL Number MFCD00005112
InChI Key NLMDJJTUQPXZFG-UHFFFAOYSA-P
Synonym kryptofix 22, cryptand 2.2, cryptand 22, diaza-18-crown-6, 1,7,10,16-tetraoxa-4,13-diazacyclooctadecane, 7,16-diaza-18-crown-6, unii-xly51t1rsz, xly51t1rsz, 1,10-diaza-4,7,13,16-tetraoxacyclooctadecane, 7,16-diaza-1,4,10,13-tetraoxcyclooctadecane
PubChem CID 72805
IUPAC Name 1,4,10,13-tetraoxa-7,16-diazacyclooctadecane-7,16-diium
SMILES C1COCCOCC[NH2+]CCOCCOCC[NH2+]1

Specifications

Melting Point 115°C
Color White-Yellow
Quantity 5 g
Formula Weight 262.35
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 4,13-Diaza-18-crown 6-Ether