accessibility menu, dialog, popup

UserName

CH3CH3H3COONH(S)NHOOOHO

CAS RN 2389-45-9

IUPAC Name: (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
Synonyms: (2S)-6-{[(benzyloxy)carbonyl]amino}-2-[(tert-butoxycarbonyl)amino]hexanoic acid
Molecular Weight (g/mol): 380.44
Molecular Formula: C19H28N2O6
InChi Key: BDHUTRNYBGWPBL-GGYSOQFKNA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)