missing translation for 'onlineSavingsMsg'
Learn More

Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine, 95%

Catalog No. AAH5953709
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5953709 10 g
AAH5953704 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5953709 Supplier Thermo Scientific Chemicals Supplier No. H5953709
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2389-45-9 | C19H28N2O6 | 380.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2389-45-9
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00065584
InChI Key BDHUTRNYBGWPBL-GGYSOQFKNA-N
Synonym boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
PubChem CID 2724765
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Quantity 10 g
Beilstein 1917222
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-6-(phenylmethoxycarbonylamino)hexanoic acid
Formula Weight 380.44
Percent Purity 95%
Chemical Name or Material Nepsilon-Benzyloxycarbonyl-Nalpha-Boc-L-lysine
EINECSNumber 219-221-0
TSCA Yes
Recommended Storage Store at -20°C

RUO – Research Use Only