missing translation for 'onlineSavingsMsg'
Learn More

Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine 98.0+%, TCI America™

Catalog No. B16325G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B16325G 5 g
B163225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B16325G Supplier TCI America Supplier No. B16325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2389-45-9
Molecular Formula C19H28N2O6
Molecular Weight (g/mol) 380.44
MDL Number MFCD00065584
InChI Key BDHUTRNYBGWPBL-GGYSOQFKNA-N
Synonym boc-lys z-oh, n-boc-n'-cbz-l-lysine, boc-lys cbz-oh, boc-l-lys z-oh, nalpha-boc-nepsilon-cbz-l-lysine, s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, 2s-6-benzyloxy carbonyl amino-2-tert-butoxycarbonyl amino hexanoic acid, n-alpha-boc-n-epsilon-benzyloxycarbonyl-l-lysine, nalpha-tert-butoxycarbonyl-nepsilon-carbobenzoxy-l-lysine, n-alpha-t-butyloxycarbonyl-n-epsilon-benzyloxycarbonyl-l-lysine
PubChem CID 2724765
IUPAC Name (2S)-6-{[(benzyloxy)carbonyl]amino}-2-{[(tert-butoxy)carbonyl]amino}hexanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 77°C
Color White
Quantity 5 g
Formula Weight 380.44
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Nalpha-(tert-Butoxycarbonyl)-Nepsilon-carbobenzoxy-L-lysine