accessibility menu, dialog, popup

UserName

OONOHNOH(E/Z)

CAS RN 23789-34-6

IUPAC Name: N-[1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Synonyms: N-[1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
Molecular Weight (g/mol): 220.18
Molecular Formula: C10H8N2O4
InChi Key: BRKZOASOLMUAQJ-UHFFFAOYSA-N
SMILES: ONC(=C(N=O)C1=CC=CO1)C1=CC=CO1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results