missing translation for 'onlineSavingsMsg'
Learn More

alpha-Furil Dioxime 97.0+%, TCI America™

Catalog No. F00795G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F00795G 5 g
F00791G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F00795G Supplier TCI America Supplier No. F00795G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 23789-34-6
Molecular Formula C10H8N2O4
Molecular Weight (g/mol) 220.18
MDL Number MFCD00075315,MFCD00075315
InChI Key BRKZOASOLMUAQJ-UHFFFAOYSA-N
PubChem CID 5359431
IUPAC Name N-[1,2-bis(furan-2-yl)-2-nitrosoethenyl]hydroxylamine
SMILES ONC(=C(N=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 192°C
Quantity 5 g
Formula Weight 220.18
Percent Purity ≥97.0% (W)
Physical Form Crystalline Powder
Chemical Name or Material alpha-Furil Dioxime