accessibility menu, dialog, popup

UserName

FFNFNF

CAS RN 2377-81-3

IUPAC Name: tetrafluorobenzene-1,3-dicarbonitrile
Synonyms: tetrafluorobenzene-1,3-dicarbonitrile
Molecular Weight (g/mol): 200.1
Molecular Formula: C8F4N2
InChi Key: WVHMPQKZPHOCRD-UHFFFAOYSA-N
SMILES: FC1=C(F)C(C#N)=C(F)C(C#N)=C1F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results