accessibility menu, dialog, popup

UserName

BSHHOOH

CAS RN 237429-33-3

IUPAC Name: (4-sulfanylphenyl)boronic acid
Synonyms: 4-sulfanylphenylboronic acid
Molecular Weight (g/mol): 153.99
Molecular Formula: C6H7BO2S
InChi Key: AUVSUPMVIZXUOG-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(S)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results