missing translation for 'onlineSavingsMsg'
Learn More

4-Mercaptophenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. M24181G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24181G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M24181G Supplier TCI America Supplier No. M24181G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 4-Mercaptophenylboronic Acid (contains varying amounts of Anhydride)
CAS 237429-33-3
Melting Point 220°C
Color White-Yellow
Molecular Formula C6H7BO2S
Quantity 1 g
MDL Number MFCD03093887
Synonym 4-Mercaptobenzeneboronic Acid
InChI Key AUVSUPMVIZXUOG-UHFFFAOYSA-N
SMILES B(C1=CC=C(C=C1)S)(O)O
IUPAC Name (4-sulfanylphenyl)boronic acid
Molecular Weight (g/mol) 153.99
PubChem CID 4431894
Formula Weight 153.99
Physical Form Crystalline Powder
Show More Show Less