accessibility menu, dialog, popup

UserName

NH2NOHO

CAS RN 23628-31-1

IUPAC Name: 6-aminopyridine-2-carboxylic acid
Synonyms: 6-aminopyridine-2-carboxylic acid
Molecular Weight (g/mol): 138.13
Molecular Formula: C6H6N2O2
InChi Key: NMCKJFCJIHCHIS-UHFFFAOYSA-N
SMILES: NC1=CC=CC(=N1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)