missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Amino-2-pyridinecarboxylic Acid 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 315°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 138.13 |
| Percent Purity | ≥98.0% (HPLC,T) |
| Grade | Not Available |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 6-Amino-2-pyridinecarboxylic Acid |
Safety and Handling
| TSCA | No |