accessibility menu, dialog, popup

UserName

H3COON

CAS RN 23597-82-2

IUPAC Name: hexyl pyridine-3-carboxylate
Synonyms: hexyl nicotinate N-hexyl nicotinoate
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: RVYGVBZGSFLJKH-UHFFFAOYSA-N
SMILES: CCCCCCOC(=O)C1=CC=CN=C1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)