missing translation for 'onlineSavingsMsg'
Learn More

Hexyl nicotinate, 98%

Catalog No. AC460090250
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC460090250 25 g
AC460090050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC460090250 Supplier Thermo Scientific Chemicals Supplier No. 460090250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 23597-82-2 | C12H17NO2 | 207.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23597-82-2
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.27
InChI Key RVYGVBZGSFLJKH-UHFFFAOYSA-N
Synonym hexyl nicotinate, 3-pyridinecarboxylic acid, hexyl ester, hexyl nicotinoate, nicotherm, n-hexyl nicotinoate, nicotinic acid, hexyl ester, unii-bn07pb44iv, nicotinic acid n-hexyl ester, bn07pb44iv, nicotinsaeure-n-hexylester
PubChem CID 90202
IUPAC Name hexyl pyridine-3-carboxylate
SMILES CCCCCCOC(=O)C1=CN=CC=C1

Specifications

Density 1.0200g/mL
Boiling Point 329.0°C
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Specific Gravity 1.02
Formula Weight 207.27
Percent Purity 98%
Chemical Name or Material Hexyl nicotinate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation. Causes serious eye irritation.
Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
EINECSNumber 245-767-4

RUO – Research Use Only