accessibility menu, dialog, popup

UserName

CAS RN 23411-34-9

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): 394.3
Molecular Formula: C10H16CaN2Na2O9
InChi Key: WJEKUTCBXSHIOX-UHFFFAOYSA-J
SMILES: O.[Na+].[Na+].[Ca+4].[O-]C(=O)C[NH](CC[NH](CC([O-])=O)CC([O-])=O)CC([O-])=O
Grade 
  • (1)
  • (4)
Percent Purity 
  • (4)
Physical Form 
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (4)
  • (1)
  • (4)
  • (1)
  • (4)
  • (4)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)