missing translation for 'onlineSavingsMsg'
Learn More

Edetate Calcium Disodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013624
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013624 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013624 Supplier Merck Emd Millipore Supplier No. PHR15101G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Edetate Calcium Disodium is a metal-chelating agent, typically used for the treatment of symptomatic and severe lead poisoning. It also finds application in the prevention of calcium depletion in the body. It can be used as a flavoring and color retention agent in food products.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 23411-34-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula Ca(OOCCH2)2 NCH2 CH2 N(CH2 COONa)2 · aq
MDL Number MFCD00149453
Synonym Ethylenediaminetetraacetic acid calcium disodium salt hydrate; Calcium disodium ethylenediaminetetraacetate hydrate
Type Certified Reference Material
Quantity 1 g
Formula Weight 374.27 (anhydrous basis)
Grade Certified Reference Material