accessibility menu, dialog, popup

UserName

HO(S)(S)OH

CAS RN 2325-10-2

IUPAC Name: (1S,2S)-1,2-diphenylethane-1,2-diol
Synonyms: (1S,2S)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol): 214.26
Molecular Formula: C14H14O2
InChi Key: IHPDTPWNFBQHEB-KBPBESRZSA-N
SMILES: O[C@H]([C@@H](O)C1=CC=CC=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)