missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-Hydrobenzoin 99.0+%, TCI America™

Catalog No. H08245G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H08245G 5 g
H082425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H08245G Supplier TCI America Supplier No. H08245G

Chemical Identifiers

CAS 2325-10-2
Molecular Formula C14H14O2
Molecular Weight (g/mol) 214.26
MDL Number MFCD00064255
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
PubChem CID 853020
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O

Specifications

Melting Point 148°C
Color White
Quantity 5 g
Formula Weight 214.26
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S,S)-(-)-Hydrobenzoin