missing translation for 'onlineSavingsMsg'
Learn More

(S,S)-(-)-Hydrobenzoin, 98+%

Catalog No. AAL1408214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1408214 25 g
AAL1408206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1408214 Supplier Thermo Scientific Chemicals Supplier No. L1408214

CAS: 2325-10-2 | C14H14O2 | 214.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2325-10-2
MDL Number MFCD00064255

Specifications

Melting Point 148°C to 150°C
Color White
Molecular Formula C14H14O2
Quantity 25 g
Beilstein 2330888
Merck Index 14,4777
Synonym s,s---hydrobenzoin, 1s,2s-1,2-diphenylethane-1,2-diol, s,s-hydrobenzoin, s,s---1,2-diphenyl-1,2-ethanediol, hydrobenzoin,-, unii-wx45q7714b, 1,2-ethanediol, 1,2-diphenyl-, 1s,2s, s,s-1,2-diphenylethylene glycol, 1s,2s---1,2-diphenyl-1,2-ethanediol, isohydrobenzoin
InChI Key IHPDTPWNFBQHEB-KBPBESRZSA-N
SMILES C1=CC=C(C=C1)C(C(C2=CC=CC=C2)O)O
IUPAC Name (1S,2S)-1,2-diphenylethane-1,2-diol
Molecular Weight (g/mol) 214.26
PubChem CID 853020
Formula Weight 214.27
Percent Purity ≥98%
Physical Form Crystals
Chemical Name or Material (S,S)-(-)-Hydrobenzoin
Show More Show Less
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only