accessibility menu, dialog, popup

UserName

CAS RN 2312-15-4

IUPAC Name: 4-(dodecyloxy)benzoic acid
Synonyms: benzoic acid, 4-(dodecyloxy)-
Molecular Weight (g/mol): 306.45
Molecular Formula: C19H30O3
InChi Key: ALQLYJHDBAKLBB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC1=CC=C(C=C1)C(O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)