accessibility menu, dialog, popup

UserName

CH3OOCH3BOHHO

CAS RN 23112-96-1

IUPAC Name: (2,6-dimethoxyphenyl)boronic acid
Synonyms: 2,6-dimethoxyphenylboronic acid
Molecular Weight (g/mol): 181.98
Molecular Formula: C8H11BO4
InChi Key: BKWVXPCYDRURMK-UHFFFAOYSA-N
SMILES: COC1=CC=CC(OC)=C1B(O)O
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)