missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. D38615G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D38615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D38615G Supplier TCI America Supplier No. D38615G

Chemical Identifiers

CAS 23112-96-1
Molecular Formula C8H11BO4
MDL Number MFCD01318987
InChI Key BKWVXPCYDRURMK-UHFFFAOYSA-N
PubChem CID 2734343
IUPAC Name (2,6-dimethoxyphenyl)boronic acid

Specifications

Melting Point 113°C
Color White-Yellow
Quantity 5 g
Synonym 2,6-dimethoxyphenyl boronic acid, 2,6-dimethoxyphenylboronicacid, 2,6-dimethoxybenzeneboronic acid, 2,6-dimethoxyphenyl-boronic acid, 2,6-dimethoxyphenyl boranediol, boronic acid, 2,6-dimethoxyphenyl, pubchem1825, acmc-1clhv, ksc201q0f, 2,6-dimethoxy-phenylboronic acid
SMILES B(C1=C(C=CC=C1OC)OC)(O)O
Molecular Weight (g/mol) 181.982
Formula Weight 181.98
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Dimethoxyphenylboronic Acid (contains varying amounts of Anhydride)