accessibility menu, dialog, popup

UserName

CH3NNH3CH3CFPFFFFF

CAS RN 227617-70-1

IUPAC Name: 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
Synonyms: 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate
Molecular Weight (g/mol): 298.21
Molecular Formula: C9H17F6N2P
InChi Key: JWFPQAXAGSAKRF-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)