missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium Hexafluorophosphate 97.0+%, TCI America™

Catalog No. B24745G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B24745G 5 g
B247425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B24745G Supplier TCI America Supplier No. B24745G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 227617-70-1
Molecular Formula C9H17F6N2P
Molecular Weight (g/mol) 298.21
MDL Number MFCD03790877
InChI Key JWFPQAXAGSAKRF-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636
PubChem CID 11243457
IUPAC Name 1-butyl-2,3-dimethyl-1H-imidazol-3-ium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 298.21
Percent Purity ≥97.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium Hexafluorophosphate