missing translation for 'onlineSavingsMsg'
Learn More

1-Butyl-2,3-dimethylimidazolium hexafluorophosphate, 99%

Catalog No. AAH2782718
Change view
Click to view available options
Quantity:
5 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2782718 50 g
AAH2782706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2782718 Supplier Thermo Scientific Chemicals Supplier No. H2782718

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 227617-70-1 | C9H17F6N2P | 298.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 227617-70-1
Molecular Formula C9H17F6N2P
Molecular Weight (g/mol) 298.21
MDL Number MFCD03790877
InChI Key JWFPQAXAGSAKRF-UHFFFAOYSA-N
Synonym 1-butyl-2,3-dimethylimidazolium hexafluorophosphate, 1-butyl-2,3-dimethyl-1h-imidazol-3-ium hexafluorophosphate v, 1-butyl-2,3-dimethylimidazol-1-ium hexafluorophosphate, 3-butyl-1,2-dimethylimidazol-1-ium hexafluorophosphate, acmc-209fyn, bdmim pf6, c4c1mim pf6, c4mmim pf6, dsstox_cid_27883, dsstox_rid_82636
PubChem CID 11243457
IUPAC Name 1-butyl-2,3-dimethylimidazol-3-ium;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.CCCCN1C=C[N+](C)=C1C

Specifications

Melting Point 48°C to 50°C
Density 1.354
Refractive Index 1.423
Quantity 50 g
Formula Weight 298.21
Percent Purity 99%
Chemical Name or Material 1-Butyl-2,3-dimethylimidazolium hexafluorophosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only