accessibility menu, dialog, popup

UserName

CH3SOON

CAS RN 2274-42-2

IUPAC Name: 2-methanesulfonylacetonitrile
Synonyms: methanesulfonylacetonitrile
Molecular Weight (g/mol): 119.14
Molecular Formula: C3H5NO2S
InChi Key: FOTRKCAZUSJCQD-UHFFFAOYSA-N
SMILES: CS(=O)(=O)CC#N
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)