missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylsulfonyl)acetonitrile, 97%

Catalog No. AC438420250
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC438420250 25 g Glass bottle
AC438420050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC438420250 Supplier Thermo Scientific Chemicals Supplier No. 438420250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2274-42-2 | C3H5NO2S | 119.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2274-42-2
Molecular Formula C3H5NO2S
Molecular Weight (g/mol) 119.14
InChI Key FOTRKCAZUSJCQD-UHFFFAOYSA-N
Synonym methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
PubChem CID 75283
ChEBI CHEBI:32444
IUPAC Name 2-methylsulfonylacetonitrile
SMILES CS(=O)(=O)CC#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 83.0°C to 86.0°C
Color White
Elemental Analysis 29.03 to 31.45% C, 11.29 to 12.23% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 25 g
Formula Weight 119.14
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 2-(Methylsulfonyl)acetonitrile
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
EINECSNumber 218-888-5

RUO – Research Use Only