missing translation for 'onlineSavingsMsg'
Learn More

Methylsulfonylacetonitrile 98.0+%, TCI America™

Catalog No. M06675G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M06675G 5 g
M066725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M06675G Supplier TCI America Supplier No. M06675G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2274-42-2
Molecular Formula C3H5NO2S
Molecular Weight (g/mol) 119.138
MDL Number MFCD00014743
InChI Key FOTRKCAZUSJCQD-UHFFFAOYSA-N
Synonym methylsulfonylacetonitrile, 2-methylsulfonyl acetonitrile, methylsulfonyl acetonitrile, methanesulfonylacetonitrile, mesylacetonitrile, 2-methanesulfonylacetonitrile, cyanomethylsulfonylmethane, methanesulphonylacetonitrile, acetonitrile, methylsulfonyl, cyanomethyl methyl sulphone
PubChem CID 75283
ChEBI CHEBI:32444
IUPAC Name 2-methylsulfonylacetonitrile
SMILES CS(=O)(=O)CC#N

Specifications

Melting Point 84°C
Color White-Yellow
Quantity 5 g
UN Number 3439
Formula Weight 119.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methylsulfonylacetonitrile