accessibility menu, dialog, popup

UserName

H2NOBr

CAS RN 22726-00-7

IUPAC Name: 3-bromobenzamide
Synonyms: M-bromobenzamide
Molecular Weight (g/mol): 200.04
Molecular Formula: C7H6BrNO
InChi Key: ODJFDWIECLJWSR-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=CC(Br)=C1
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)