accessibility menu, dialog, popup

UserName

CAS RN 22307-72-8

H3CNCH3H3CH3COOHO

CAS RN 22307-72-8

IUPAC Name: tetrabutylazanium 2-carboxybenzen-1-olate
Synonyms: tetrabutylammonium ion 2-carboxybenzenolate
Molecular Weight (g/mol): 379.59
Molecular Formula: C23H41NO3
InChi Key: ZWRWGKGPUFESNE-UHFFFAOYSA-M
SMILES: OC(=O)C1=CC=CC=C1[O-].CCCC[N+](CCCC)(CCCC)CCCC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results