accessibility menu, dialog, popup

UserName

CAS RN 22237-13-4

IUPAC Name: (4-ethoxyphenyl)boronic acid
Synonyms: 4-ethoxyphenylboronic acid
Molecular Weight (g/mol): 165.98
Molecular Formula: C8H11BO3
InChi Key: WRQNDLDUNQMTCL-UHFFFAOYSA-N
SMILES: CCOC1=CC=C(C=C1)B(O)O
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)