missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E07251G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07251G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07251G Supplier TCI America Supplier No. E07251G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22237-13-4
Molecular Formula C8H11BO3
MDL Number MFCD00674028
InChI Key WRQNDLDUNQMTCL-UHFFFAOYSA-N
PubChem CID 2734351
IUPAC Name (4-ethoxyphenyl)boronic acid

Specifications

Melting Point 168°C
Color White-Yellow
Quantity 1 g
Synonym 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
SMILES B(C1=CC=C(C=C1)OCC)(O)O
Molecular Weight (g/mol) 165.983
Formula Weight 165.98
Physical Form Crystalline Powder
Chemical Name or Material 4-Ethoxyphenylboronic Acid (contains varying amounts of Anhydride)