missing translation for 'onlineSavingsMsg'
Learn More

4-Ethoxybenzeneboronic acid, 98%

Catalog No. AAB2368314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2368314 25 g
AAB2368303 1 g
AAB2368306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2368314 Supplier Thermo Scientific Chemicals Supplier No. B2368314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 22237-13-4 | C8H11BO3 | 165.983 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 22237-13-4
Molecular Formula C8H11BO3
Molecular Weight (g/mol) 165.983
MDL Number MFCD00674028
InChI Key WRQNDLDUNQMTCL-UHFFFAOYSA-N
Synonym 4-ethoxybenzeneboronic acid, 4-ethoxyphenyl boronic acid, 4-ethoxyphenyl boranediol, p-ethoxyphenylboronic acid, 4-n-ethoxyphenyl boronic acid, boronic acid, 4-ethoxyphenyl, 4-ethoxybenzeneboronicacid, 4-ethoxyphenylboronicacid, 4-ethoxyphenylboronic
PubChem CID 2734351
IUPAC Name (4-ethoxyphenyl)boronic acid
SMILES B(C1=CC=C(C=C1)OCC)(O)O

Specifications

Melting Point 165°C to 168°C
Quantity 25 g
Beilstein 3530408
Formula Weight 165.99
Percent Purity 98%
Chemical Name or Material 4-Ethoxybenzeneboronic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only