missing translation for 'onlineSavingsMsg'
Learn More

Carbimazole Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013575
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013575 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013575 Supplier Merck Emd Millipore Supplier No. PHR16841G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Carbimazole is an orally administered thioimidazole drug, which is indicated for the management of hyperthyroidism as well as Grave′s disease in humans. It is used to facilitate the growth of animals for human consumption.Pharmaceutical secondary standard for application in quality control. Provides pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 22232-54-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C7H10N2O2 S
Synonym Carbimazole; 2,3-Dihydro-3-methyl-2-thioxo-1 H-imidazole-1-carboxylic acid ethyl ester
Type Certified Reference Material
Quantity 1 g
Formula Weight 186.23
Grade Certified Reference Material