accessibility menu, dialog, popup

UserName

H3COONH

CAS RN 2199-43-1

IUPAC Name: ethyl 1H-pyrrole-2-carboxylate
Synonyms: ethyl 1H-pyrrole-2-carboxylate
Molecular Weight (g/mol): 139.15
Molecular Formula: C7H9NO2
InChi Key: PAEYAKGINDQUCT-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CN1
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)