missing translation for 'onlineSavingsMsg'
Learn More

Ethyl pyrrole-2-carboxylate, 98+%

Catalog No. AAL1638214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1638214 25 g
AAL1638206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1638214 Supplier Thermo Scientific Chemicals Supplier No. L1638214
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2199-43-1 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2199-43-1
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00817049
InChI Key PAEYAKGINDQUCT-UHFFFAOYSA-N
Synonym ethyl pyrrole-2-carboxylate, ethyl 2-pyrrolecarboxylate, ethyl pyrrole-2-carbonate, ethylpyrrole-2-carboxylate, 1h-pyrrole-2-carboxylic acid, ethyl ester, 2-ethoxycarbonyl-1h-pyrrole, pyrrole-2-carboxylic acid, ethyl ester, 1h-pyrrole-2-carboxylic acid ethyl ester, pyrrole-2-carboxylic acid ethyl ester, ethyl1h-pyrrole-2-carboxylate
PubChem CID 255670
IUPAC Name ethyl 1H-pyrrole-2-carboxylate
SMILES CCOC(=O)C1=CC=CN1

Specifications

Melting Point 37°C to 39°C
Boiling Point 70°C (0.8 mmHg)
Quantity 25 g
Beilstein 2645
Formula Weight 139.16
Percent Purity ≥98%
Chemical Name or Material Ethyl pyrrole-2-carboxylate
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only